Merck
  • Tandem Blaise-alkenylation with unactivated alkynes: one-pot synthesis of alpha-vinylated beta-enaminoesters from nitriles.
Tandem Blaise-alkenylation with unactivated alkynes: one-pot synthesis of alpha-vinylated beta-enaminoesters from nitriles.
Organic letters (2009-07-04)
Yu Sung Chun, Young Ok Ko, Hyunik Shin, Sang-gi Lee
ABSTRACT
The in situ generated Blaise reaction intermediate, a zinc bromide complex of beta-enaminoester, reacts with various unactivated terminal alkynes and an internal alkyne under mild conditions to afford alpha-vinylated beta-enaminoesters in good to excellent yields.
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, puriss., anhydrous, ≥98%