Merck
  • Direct electrochemistry and spectroelectrochemistry of osmium substituted horseradish peroxidase.
Direct electrochemistry and spectroelectrochemistry of osmium substituted horseradish peroxidase.
Bioelectrochemistry (Amsterdam, Netherlands) (2009-05-05)
Noya Loew, Ulla Wollenberger, Frieder W Scheller, Martin Katterle
ABSTRACT
In this contribution the substitution of the central protoporphyrin IX iron complex of horseradish peroxidase by the respective osmium porphyrin complex is described. The direct electrochemical reduction of the Os containing horseradish peroxidase (OsHRP) was achieved at ITO and modified glassy carbon electrodes and in combination with spectroscopy revealed the three redox couples Os(III)HRP/Os(IV)HRP, Os(IV)HRP/Os(V)HRP and Os(V)HRP/Os(VI)HRP. The midpoint potentials differ dependent on the electrode material used with E(1/2) (Os(III/IV)) of -0.4 V (ITO) and -0.25 V (GC), E(1/2) (Os(IV)/(V)) of -0.16 V (ITO) and +0.10 V (GC), and E(1/2) (Os(V/VI))of +0.18 V (ITO), respectively. Moreover, with immobilised OsHRP the direct electrocatalytic reduction of hydrogen peroxide and tert-butyl hydroperoxide was observed. In comparison to electrodes modified with native HRP the sensitivity of the OsHRP-electrode for tert-butyl hydroperoxide is higher.
Sigma-Aldrich
Triirondodecacarbonyl, contains 1-10% methyl alcohol
Sigma-Aldrich
Protoporphyrin IX dimethyl ester, ~90% (HPLC)