Merck
  • 2-Acetylfuran, a confounder in urinalysis for 2,5-hexanedione as an n-hexane exposure indicator.
2-Acetylfuran, a confounder in urinalysis for 2,5-hexanedione as an n-hexane exposure indicator.
International archives of occupational and environmental health (1991-01-01)
T Kawai, T Yasugi, K Mizunuma, S Horiguchi, Y Uchida, O Iwami, H Iguchi, M Ikeda
ABSTRACT
The apparent amount of 2,5-hexanedione, a biomarker of n-hexane expsoure in occupational health, in the urine of both exposed and non-exposed subjects varied not only as a function of the pH at which the urine sample was hydrolyzed but also depending on the capillary column used for gas chromatographic (GC) analysis of the urinary hydrolyzates after extraction with dichloromethane. The formation of a compound, identified by gas chromatography-mass spectrometry (GC-MS) as 2-acetylfuran, following acid hydrolysis was a major cause of confounding effects. This compound was hardly separated from 2.5-hexanedione on a capillary column such as DB-WAX, whereas separation could be achieved on a DB-1 capillary column. 2-Acetylfuran was formed when a urine sample was heated at a pH of less than 2 for hydrolysis, and the amount detected in urine did not differ between exposed and non-exposed subjects, indicating that the formation of 2-acetylfuran is independent of n-hexane exposure. When urinary hydrolysis is used, hydrolysis at a pH of less than 0.5, extraction with dichloromethane, and GC analysis on a non-polar capillary column are proposed to be the best analytical conditions for 2,5-hexanedione analysis in biological monitoring of exposure to n-hexane.
Sigma-Aldrich
2-Furyl methyl ketone, 99%
Sigma-Aldrich
2-Furyl methyl ketone, ≥99%, FG
Sigma-Aldrich
2-Furyl methyl ketone, natural (US), ≥97%, FG