Merck
  • Preparation of (Cyanomethylene)tributylphosphorane: a new mitsunobu-type reagent.
Preparation of (Cyanomethylene)tributylphosphorane: a new mitsunobu-type reagent.
Chemical & pharmaceutical bulletin (2005-11-08)
Izumi Sakamoto, Takeshi Nishii, Fumie Ozaki, Hiroto Kaku, Masami Tanaka, Tetsuto Tsunoda
ABSTRACT
(Cyanomethylene)tributylphosphorane (CMBP), which promoted the alkylation of various nucleophiles (HA) with alcohols (ROH) to give RA (Mitsunobu-type reaction), was prepared in two steps starting from chloroacetonitrile.
Sigma-Aldrich
(Tributylphosphoranylidene)acetonitrile solution, 1 M in toluene
Sigma-Aldrich
Chloroacetonitrile, 99%