Merck
  • Phosphorylation-dependent mitotic SUMOylation drives nuclear envelope-chromatin interactions.
Phosphorylation-dependent mitotic SUMOylation drives nuclear envelope-chromatin interactions.
The Journal of cell biology (2021-11-18)
Christopher Ptak, Natasha O Saik, Ashwini Premashankar, Diego L Lapetina, John D Aitchison, Ben Montpetit, Richard W Wozniak
ABSTRACT
In eukaryotes, chromatin binding to the inner nuclear membrane (INM) and nuclear pore complexes (NPCs) contributes to spatial organization of the genome and epigenetic programs important for gene expression. In mitosis, chromatin-nuclear envelope (NE) interactions are lost and then formed again as sister chromosomes segregate to postmitotic nuclei. Investigating these processes in S. cerevisiae, we identified temporally and spatially controlled phosphorylation-dependent SUMOylation events that positively regulate postmetaphase chromatin association with the NE. Our work establishes a phosphorylation-mediated targeting mechanism of the SUMO ligase Siz2 to the INM during mitosis, where Siz2 binds to and SUMOylates the VAP protein Scs2. The recruitment of Siz2 through Scs2 is further responsible for a wave of SUMOylation along the INM that supports the assembly and anchorage of subtelomeric chromatin at the INM and localization of an active gene (INO1) to NPCs during the later stages of mitosis and into G1-phase.
Sigma-Aldrich
α1-Mating Factor acetate salt, ≥93% (HPLC), synthetic
Sigma-Aldrich
Anti-Protein A antibody produced in rabbit, fractionated antiserum, lyophilized powder