Merck
  • Biodegradable chelating agent improves the survival of early larvae for shellfish aquaculture.
Biodegradable chelating agent improves the survival of early larvae for shellfish aquaculture.
Aquatic toxicology (Amsterdam, Netherlands) (2020-10-04)
Daniel R McDougall, Shinji Kihara, Juliane Reinhardt, Gordon M Miskelly, Duncan J McGillivray, Andrew G Jeffs
ABSTRACT
The toxicity of heavy metals commonly impacts the survival of crustacean and bivalve larvae in hatchery culture, and this has led to the widespread use of EDTA to decrease this toxicity. Since EDTA has a very poor biodegradability leading to potential persistent environmental effects, alternative methods to prevent heavy metal toxicity to shellfish larvae are needed. EDDS is a biodegradable potential alternative to EDTA for this application and was tested as a treatment of the seawater used for rearing aquaculture Greenshell™ mussel (Perna canaliculus) larval embryos in this study. Mussel embryos reared with EDTA or EDDS had significantly better survival than without. The concentrations and spatial distributions of heavy metals in D-veliger larvae as determined by Inductively Coupled Plasma Mass Spectrometry (ICP-MS) and X-ray Fluorescence Microscopy (XFM) suggested that chelating agents increased the levels of calcium in larvae while they reduced the concentration of zinc. In addition, where decreased accumulation of the other heavy metals was not observed, chelating agents affected their distribution within the larvae, especially for copper and arsenic. This is the first study to test the use of EDDS for aquaculture hatchery application and shows that EDDS is an effective biodegradable alternative to EDTA that can mitigate the effects of heavy metals for shellfish larval rearing.
Sigma-Aldrich
Ethylenediamine-N,N′-disuccinic acid trisodium salt solution, ~35% in H2O