Merck
Effects of Gentiana lutea Root on Vascular Diseases.
Current vascular pharmacology (2020-05-30)
Gordana Joksic, Djordje Radak, Emina Sudar-Milovanovic, Milan Obradovic, Jelena Radovanovic, Esma R Isenovic
ABSTRACT
Gentiana lutea (GL), commonly known as yellow gentian, bitter root, and bitterwort, belongs to family Gentianaceae. GL belongs to genus Gentiana, which is a rich natural source of iridoids, secoiridoids, xantones, flavonoids, triterpenoids, and carbohydrates. Medicinal plants from Gentiana species have anti-oxidant, anti-inflammatory, anti-mitogenic, anti-proliferative, and lipidlowering effects, as well as a cardioprotective, hypotensive, vasodilator and anti-platelet activities. We reviewed the recent literature related to the effects of Gentiana species, and their active components on vascular diseases. Data used for this review were obtained by searching the electronic database [PUBMED/ MEDLINE 1973 - February 2020]. The primary data search terms of interest were: Gentiana lutea, Gentienacea family, phytochemistry, vascular diseases, treatment of vascular diseases, antioxidant, anti-inflammatory, anti-atherogenic. Gentiana species and their constituents affect many different factors related to vascular disease development and progression. Therefore, Gentiana-based therapeutics represent potentially useful drugs for the management of vascular diseases.
Product Description
Amarogentin, phyproof® Reference Substance
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %