Merck
  • Human ribonuclease 1 serves as a secretory ligand of ephrin A4 receptor and induces breast tumor initiation.
Human ribonuclease 1 serves as a secretory ligand of ephrin A4 receptor and induces breast tumor initiation.
Nature communications (2021-05-15)
Heng-Huan Lee, Ying-Nai Wang, Wen-Hao Yang, Weiya Xia, Yongkun Wei, Li-Chuan Chan, Yu-Han Wang, Zhou Jiang, Shouping Xu, Jun Yao, Yufan Qiu, Yi-Hsin Hsu, Wei-Lun Hwang, Meisi Yan, Jong-Ho Cha, Jennifer L Hsu, Jia Shen, Yuanqing Ye, Xifeng Wu, Ming-Feng Hou, Lin-Ming Tseng, Shao-Chun Wang, Mei-Ren Pan, Chin-Hua Yang, Yuan-Liang Wang, Hirohito Yamaguchi, Da Pang, Gabriel N Hortobagyi, Dihua Yu, Mien-Chie Hung
ABSTRACT
Human ribonuclease 1 (hRNase 1) is critical to extracellular RNA clearance and innate immunity to achieve homeostasis and host defense; however, whether it plays a role in cancer remains elusive. Here, we demonstrate that hRNase 1, independently of its ribonucleolytic activity, enriches the stem-like cell population and enhances the tumor-initiating ability of breast cancer cells. Specifically, secretory hRNase 1 binds to and activates the tyrosine kinase receptor ephrin A4 (EphA4) signaling to promote breast tumor initiation in an autocrine/paracrine manner, which is distinct from the classical EphA4-ephrin juxtacrine signaling through contact-dependent cell-cell communication. In addition, analysis of human breast tumor tissue microarrays reveals a positive correlation between hRNase 1, EphA4 activation, and stem cell marker CD133. Notably, high hRNase 1 level in plasma samples is positively associated with EphA4 activation in tumor tissues from breast cancer patients, highlighting the pathological relevance of the hRNase 1-EphA4 axis in breast cancer. The discovery of hRNase 1 as a secretory ligand of EphA4 that enhances breast cancer stemness suggests a potential treatment strategy by inactivating the hRNase 1-EphA4 axis.
Sigma-Aldrich
Anti-RNASE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-EPHA4 Mouse mAb (6H7), liquid, clone 6H7, Calbiochem®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit