Merck
  • The PARK2 Mutation Associated with Parkinson's Disease Enhances the Vulnerability of Peripheral Blood Lymphocytes to Paraquat.
The PARK2 Mutation Associated with Parkinson's Disease Enhances the Vulnerability of Peripheral Blood Lymphocytes to Paraquat.
BioMed research international (2020-10-09)
Fengyu Ming, Jieqiong Tan, Lixia Qin, Hainan Zhang, Jianguang Tang, Xuling Tan, Chunyu Wang
ABSTRACT
Parkinson's disease (PD) is the second most common neurodegenerative disease in middle-aged and elderly people. However, the etiology and pathogenesis of PD are still unclear and there is a lack of reliable biomarkers for early molecular diagnosis. Parkin (encoded by PARK2) is a ubiquitin E3 ligase that participates in mitochondrial homeostasis, the ubiquitin-proteasome pathway, oxidative stress response, and cell death pathways, which are involved in the pathogenesis of PD. However, Parkin is also expressed in peripheral blood lymphocytes (PBLs). In this study, permanent lymphocyte lines were established from the peripheral blood of sporadic PD (sPD) patients, PARK2 mutation carriers, and healthy controls. Reactive oxygen species (ROS), function of the mitochondrial respiratory chain complex I, and apoptosis were analyzed in the PBLs. There was no significant difference in ROS, mitochondrial respiratory chain complex I, and apoptosis between the experimental groups and the control group without paraquat treatment. Compared with the control group of healthy subjects, we found an increase of ROS (control 100 ± 0, sPD 275.53 ± 79.11, and C441R 340 ± 99.67) and apoptosis, as well as a decline in the function of mitochondrial respiratory chain complex I in PBLs of PARK2 mutation carriers and sPD after the treatment of paraquat (control 0.65 ± 0.08, sPD 0.44 ± 0.08, and C441R 0.32 ± 0.08). Moreover, overexpression of the wild-type (WT) PARK2 in HeLa cells and immortalized PBLs could rescue mitochondrial function and partially inhibit apoptosis following paraquat treatment, while the C441R mutation could not. Thus, ROS levels, activity of mitochondrial respiratory chain complex I, and apoptosis of PBLs are potential diagnostic biomarkers of PD.
Sigma-Aldrich
Annexin V-FITC Apoptosis Detection Kit
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Lymphocyte Separation Medium 1077, 500 ml