Merck
  • Vaccine-elicited 10-kilodalton culture filtrate protein-specific CD8+ T cells are sufficient to mediate protection against Mycobacterium tuberculosis infection.
Vaccine-elicited 10-kilodalton culture filtrate protein-specific CD8+ T cells are sufficient to mediate protection against Mycobacterium tuberculosis infection.
Infection and immunity (2008-03-12)
Ying Wu, Joshua S Woodworth, Daniel S Shin, Sheldon Morris, Samuel M Behar
ABSTRACT
The 10-kDa culture filtrate protein (CFP-10) and 6-kDa early secretory antigen of T cells (ESAT-6) are secreted in abundance by Mycobacterium tuberculosis and are frequently recognized by T cells from infected people. The genes encoding these proteins have been deleted from the genome of the vaccine strain Mycobacterium bovis bacillus Calmette-Guérin (BCG), and it is hypothesized that these proteins are important targets of protective immunity. Indeed, vaccination with ESAT-6 elicits protective CD4+ T cells in C57BL/6 mice. We have previously shown that M. tuberculosis infection of C3H mice elicits CFP-10-specific CD8+ and CD4+ T cells. Here we demonstrate that immunization with a CFP-10 DNA vaccine stimulates a specific T-cell response only to the H-2K(k)-restricted epitope CFP-10(32-39). These CFP-10(32-39)-specific CD8+ cells undergo a rapid expansion and accumulate in the lung following challenge of immunized mice with aerosolized M. tuberculosis. Protective immunity is induced by CFP-10 DNA vaccination as measured by a CFU reduction in the lung and spleen 4 and 8 weeks after challenge with M. tuberculosis. These data demonstrate that CFP-10 is a protective antigen and that CFP-10(32-39)-specific CD8+ T cells elicited by vaccination are sufficient to mediate protection against tuberculosis.
Sigma-Aldrich
NBT-BCIP® solution, BioReagent
Pricing and availability is not currently available.