Merck
  • Disruption of the Jak1 gene demonstrates obligatory and nonredundant roles of the Jaks in cytokine-induced biologic responses.
Disruption of the Jak1 gene demonstrates obligatory and nonredundant roles of the Jaks in cytokine-induced biologic responses.
Cell (1998-05-20)
S J Rodig, M A Meraz, J M White, P A Lampe, J K Riley, C D Arthur, K L King, K C Sheehan, L Yin, D Pennica, E M Johnson, R D Schreiber
ABSTRACT
Herein we report the generation of mice lacking the ubiquitously expressed Janus kinase, Jak1. Jak1-/- mice are runted at birth, fail to nurse, and die perinatally. Although Jak1-/- cells are responsive to many cytokines, they fail to manifest biologic responses to cytokines that bind to three distinct families of cytokine receptors. These include all class II cytokine receptors, cytokine receptors that utilize the gamma(c) subunit for signaling, and the family of cytokine receptors that depend on the gp130 subunit for signaling. Our results thus demonstrate that Jak1 plays an essential and nonredundant role in promoting biologic responses induced by a select subset of cytokine receptors, including those in which Jak utilization was thought to be nonspecific.
Sigma-Aldrich
Jak1 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)