Merck
  • Nuclear FAK promotes cell proliferation and survival through FERM-enhanced p53 degradation.
Nuclear FAK promotes cell proliferation and survival through FERM-enhanced p53 degradation.
Molecular cell (2008-01-22)
Ssang-Taek Lim, Xiao Lei Chen, Yangmi Lim, Dan A Hanson, Thanh-Trang Vo, Kyle Howerton, Nicholas Larocque, Susan J Fisher, David D Schlaepfer, Dusko Ilic
ABSTRACT
FAK is known as an integrin- and growth factor-associated tyrosine kinase promoting cell motility. Here we show that, during mouse development, FAK inactivation results in p53- and p21-dependent mesodermal cell growth arrest. Reconstitution of primary FAK-/-p21-/- fibroblasts revealed that FAK, in a kinase-independent manner, facilitates p53 turnover via enhanced Mdm2-dependent p53 ubiquitination. p53 inactivation by FAK required FAK FERM F1 lobe binding to p53, FERM F2 lobe-mediated nuclear localization, and FERM F3 lobe for connections to Mdm2 and proteasomal degradation. Staurosporine or loss of cell adhesion enhanced FERM-dependent FAK nuclear accumulation. In primary human cells, FAK knockdown raised p53-p21 levels and slowed cell proliferation but did not cause apoptosis. Notably, FAK knockdown plus cisplatin triggered p53-dependent cell apoptosis, which was rescued by either full-length FAK or FAK FERM re-expression. These studies define a scaffolding role for nuclear FAK in facilitating cell survival through enhanced p53 degradation under conditions of cellular stress.
Sigma-Aldrich
Anti-Cyclin E Antibody, Upstate®, from rabbit