Merck
NUTS and BOLTS: applications of fluorescence-detected sedimentation.
Analytical biochemistry (2008-12-24)
Rachel R Kroe, Thomas M Laue
ABSTRACT
Analytical ultracentrifugation is a widely used method for characterizing the solution behavior of macromolecules. However, the two commonly used detectors, absorbance and interference, impose some fundamental restrictions on the concentrations and complexity of the solutions that can be analyzed. The recent addition of a fluorescence detector for the XL-I analytical ultracentrifuge (AU-FDS) enables two different types of sedimentation experiments. First, the AU-FDS can detect picomolar concentrations of labeled solutes, allowing the characterization of very dilute solutions of macromolecules, applications we call normal use tracer sedimentation (NUTS). The great sensitivity of NUTS analysis allows the characterization of small quantities of materials and high-affinity interactions. Second, the AU-FDS allows characterization of trace quantities of labeled molecules in solutions containing high concentrations and complex mixtures of unlabeled molecules, applications we call biological on-line tracer sedimentation (BOLTS). The discrimination of BOLTS enables the size distribution of a labeled macromolecule to be determined in biological milieus such as cell lysates and serum. Examples that embody features of both NUTS and BOLTS applications are presented along with our observations on these applications.
Product Description
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131C
Avanti
18:1 PE CF, Avanti Research - A Croda Brand 810332C
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131P, powder
Avanti
18:1 PE CF, Avanti Research - A Croda Brand