Merck
  • Identification of Compounds in Coleus forskohlii Extract Involved in the Induction of Hepatic CYP and Fatty Liver in Mice.
Identification of Compounds in Coleus forskohlii Extract Involved in the Induction of Hepatic CYP and Fatty Liver in Mice.
Journal of oleo science (2019-10-05)
Keizo Umegaki, Kaori Yokotani, Shinsuke Marumoto, Mitsuo Miyazawa
ABSTRACT
Coleus forskohlii extract (CFE), a popular weight-loss herbal product, induces hepatic cytochrome P450 (CYP) and fatty liver in mice; however, its main bioactive ingredient, forskolin, does not show such effects. To ensure the safety of CFE as a dietary supplement, identification of the compounds implicated in the induction of hepatic CYP and fatty liver is required. In this study, we separated a crude CFE extract into 5 fractions (Fr.) by column chromatography and administered the fractions to mice for one week to assess their ability to induce CYP and fatty liver. CYP induction was detected for all fractions, indicating that many compounds may be involved in CYP induction, while fatty liver was only detected for Fr. 2. Further isolation and purification of Fr. 2 by column chromatography identified 14-deoxycoleon U as a major compound and crocetin dialdehyde as a pigment compound. An in vivo mouse study revealed that crocetin dialdehyde had no effect on the liver and, as 14-deoxycoleon U was the major compound in Fr. 2, it is likely that the active compound inducing fatty liver in CFE is 14-deoxycoleon U. These findings will facilitate the preparation of standardized safe CFE ingredients for dietary supplements.
Product Description
Supelco
Crocetin dialdehyde, analytical standard