
Zinc phthalocyanine

Synonym(s):
Phthalocyanine zinc salt
Empirical Formula (Hill Notation):
C32H16N8Zn
Products
Brand
Formula Weight
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing