trans-3-(4-Methoxybenzoyl)acrylic acid
Linear Formula:
CH3OC6H4COCH=CHCO2H
CAS Number:
Molecular Weight:
206.19
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing