
Timolol Related Compound F

Synonym(s):
3-Chloro-4-morpholin-4-yl-1,2,5-thiadiazole, 4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine
Empirical Formula (Hill Notation):
C6H8ClN3OS
CAS Number:
Molecular Weight:
205.67
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2