
Thiamazole

Synonym(s):
Methimazole, 1-Methyl-2-imidazolethiol, 2-Mercapto-1-methylimidazole
Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing