
Sodium Acetate Anhydrous

Synonym(s):
Sodium acetate, Acetic acid sodium salt
Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3