![Reserpine Standard for LC-MS analytical standard, for LC-MS](/deepweb/assets/sigmaaldrich/product/structures/153/733/52abef8a-c320-4d42-9340-12b671b833d1/640/52abef8a-c320-4d42-9340-12b671b833d1.png)
Reserpine Standard for LC-MS
![Reserpine Standard for LC-MS analytical standard, for LC-MS](/deepweb/assets/sigmaaldrich/product/structures/153/733/52abef8a-c320-4d42-9340-12b671b833d1/640/52abef8a-c320-4d42-9340-12b671b833d1.png)
Synonym(s):
Reserpine, (3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester
Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing