
Pectolinarin

Synonym(s):
Pectolinarin, 5,7-Dihydroxy 4′,6-dimethoxyflavone 7-rutinoside, Pectolinarigenin 7-rutinoside, Pectolinaroside
Empirical Formula (Hill Notation):
C29H34O15
CAS Number:
Molecular Weight:
622.57
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing