
OXONE®, monopersulfate compound

Synonym(s):
Potassium peroxymonosulfate
Linear Formula:
KHSO5 · 0.5KHSO4 · 0.5K2SO4
CAS Number:
Molecular Weight:
307.38
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing