![Nitenpyram PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/156/780/50217439-62bd-41ac-932c-a80ffad5d89a/640/50217439-62bd-41ac-932c-a80ffad5d89a.png)
Nitenpyram
![Nitenpyram PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/156/780/50217439-62bd-41ac-932c-a80ffad5d89a/640/50217439-62bd-41ac-932c-a80ffad5d89a.png)
Empirical Formula (Hill Notation):
C11H15ClN4O2
CAS Number:
Molecular Weight:
270.72
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2