Neurokinin A
Synonym(s):
α-Neurokinin, Neuromedin L, Substance K
Linear Formula:
C50H79N13O15S
CAS Number:
Molecular Weight:
1133.32
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1