
Nerolin bromelia

Synonym(s):
2-Ethoxynaphthalene, 2-naphthol ethyl ether, Ethyl 2-naphthyl ether, Neroline, beta-naphthol ethyl ether, beta-naphthyl ethyl ether, bromelia, ethyl beta-naphtholate, ethyl beta-naphthyl ether, nerolin bromelia
Empirical Formula (Hill Notation):
C12H12O
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Geographic Origin
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing