![NADPH powder, =97% (dry weight)](/deepweb/assets/sigmaaldrich/product/structures/381/486/9c382ca1-e8a3-4c3d-a476-91650af83747/640/9c382ca1-e8a3-4c3d-a476-91650af83747.png)
NADPH
![NADPH powder, =97% (dry weight)](/deepweb/assets/sigmaaldrich/product/structures/381/486/9c382ca1-e8a3-4c3d-a476-91650af83747/640/9c382ca1-e8a3-4c3d-a476-91650af83747.png)
Synonym(s):
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, β-NADPH, 2′-NADPH hydrate, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt
Empirical Formula (Hill Notation):
C21H26N7Na4O17P3 · xH2O
CAS Number:
Molecular Weight:
833.35 (anhydrous basis)
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing