
L-Aspartic acid β-(7-amido-4-methylcoumarin)

Synonym(s):
L-Aspartic acid β-(4-methyl-7-coumarinylamide), L-Aspartic acid 4-(4-methyl-7-coumarinylamide)
Empirical Formula (Hill Notation):
C14H14N2O5
CAS Number:
Molecular Weight:
290.27
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing