
Hydrocortisone Impurity F

Synonym(s):
Reichstein’s substance S, 11-Deoxy-17-hydroxycorticosterone, 11-Deoxycortisol, 17α,21-Dihydroxy-4-pregnene-3,20-dione, 17α-Hydroxycortexone, 17,21-Dihydroxy-4-pregnene-3,20-dione, 17,21-Dihydroxyprogesterone, 4-Pregnene-17α,21-diol-3,20-dione, Cortexolone, Reichstein’s Compound S
Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing