
Hyaluronic acid disaccharide ΔDiHA sodium salt

Synonym(s):
α-ΔUA-(1→3)-GlcNAc
Linear Formula:
C14H20NO11Na
CAS Number:
Molecular Weight:
401.30
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing