
Geranyl pyrophosphate lithium salt

Synonym(s):
(E)-3,7-Dimethyl-2,6-octadien-1-ol trihydrogen pyrophosphate lithium salt, (E)-3,7-Dimethyl-2,6-octadien-1-yl pyrophosphate lithium salt, GPP-Li
Empirical Formula (Hill Notation):
C10H20O7P2 · xLi+
CAS Number:
Molecular Weight:
314.21 (free acid basis)
Products
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing