
Fumaric acid-1,4-13C2,2,3-d2

Synonym(s):
Fumaric acid-1,4-13C2,2,3-d2
Linear Formula:
HOO13CCD=CD13COOH
CAS Number:
Molecular Weight:
120.07
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Special Grade
Technique
Showing 1-2 of 2