Skip to Content
Merck
vibrant-m

Fluorescein Related Compound C

vibrant-m
Synonym(s):
2-(2,4-Dihydroxybenzoyl)benzoic acid
Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1