
Fludarabine phosphate

Synonym(s):
2-Fluoro-9-(5-O-phosphono-β-D-arabinofuranosyl)-9H-purin-6-amine
Empirical Formula (Hill Notation):
C10H13FN5O7P
CAS Number:
Molecular Weight:
365.21
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3