Skip to Content
Merck
Ethyl (R)-(−)-2-hydroxy-4-phenylbutyrate 98%

Ethyl (R)-(−)-2-hydroxy-4-phenylbutyrate

Ethyl (R)-(−)-2-hydroxy-4-phenylbutyrate 98%
Linear Formula:
C6H5CH2CH2CH(OH)CO2C2H5
CAS Number:
Molecular Weight:
208.25

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing