
Efavirenz Related Compound B

Synonym(s):
(4S)-6-Chloro-4-[(E)-2-cyclopropylvinyl]-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one
Empirical Formula (Hill Notation):
C14H11ClF3NO2
CAS Number:
Molecular Weight:
317.69
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2