(±)-Vigabatrin
Synonym(s):
(R,S)-4-Amino-5-hexenoic acid, (±)-γ-Vinyl-GABA, (±)-4-Aminohexenoic acid
Empirical Formula (Hill Notation):
C6H11NO2
Products
Product Category
Brand
Formula Weight
Gene Alias
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing