
Diacerein impurity B

Synonym(s):
Aloe-emodin, 1,8-Dihydroxy 3-hydroxymethylanthraquinone, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone, 3-Hydroxymethylchrysazine, Rhabarberone
Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing