![Deferiprone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/302/931/a0c7757f-afd4-4772-ba5c-f0af8b7e79e0/640/a0c7757f-afd4-4772-ba5c-f0af8b7e79e0.png)
Deferiprone
![Deferiprone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/302/931/a0c7757f-afd4-4772-ba5c-f0af8b7e79e0/640/a0c7757f-afd4-4772-ba5c-f0af8b7e79e0.png)
Synonym(s):
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone, Deferiprone
Empirical Formula (Hill Notation):
C7H9NO2
CAS Number:
Molecular Weight:
139.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing