
Colistimethate sodium

Synonym(s):
Colistin sodium methanesulfonate, Colimycin sodium methanesulfonate, Methanesulfonic acid derivative of Polymyxin E, Polymyxin E sodium methanesulfonate
Empirical Formula (Hill Notation):
C58H115N16Na5O28S5
CAS Number:
Molecular Weight:
1759.90
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing