
Brain Heart Infusion Broth

Synonym(s):
BHI Broth, Brain Heart Broth
Products
Product Category
Brand
Color
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing