Skip to Content
Merck
Benzene-1,2-dithiol 96%

Benzene-1,2-dithiol

Benzene-1,2-dithiol 96%
Synonym(s):
1,2-Dimercaptobenzene
Linear Formula:
C6H4(SH)2
CAS Number:
Molecular Weight:
142.24
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing