![Articaine Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/409/740/9aa9633a-df23-4477-b77e-e579a83bedba/640/9aa9633a-df23-4477-b77e-e579a83bedba.jpg)
Articaine Related Compound E
![Articaine Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/409/740/9aa9633a-df23-4477-b77e-e579a83bedba/640/9aa9633a-df23-4477-b77e-e579a83bedba.jpg)
Synonym(s):
Methyl 3-[2-(isopropylamino)propanamido]-4-methyl-2-thiophenecarboxylate
Empirical Formula (Hill Notation):
C13H20N2O3S
Molecular Weight:
284.37
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2