
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–FITC antibody produced in sheep

Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing