Anti-CANX antibody produced in rabbit
Synonym(s):
Anti-Calnexin precursor antibody produced in rabbit, Anti-IP90 antibody produced in rabbit, Anti-Major histocompatibility complex class I antigen-binding protein p88 antibody produced in rabbit, Anti-p90 antibody produced in rabbit, Anti-CANX, Anti-CNX, Anti-Calnexin -NT, Anti-IP90, Anti-P90, Anti-Calnexin-CT (Purified)
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing