
Andrograpanin

Synonym(s):
3,14-Dideoxyandrographolide, 3-[2-[(1R,4aS,5R,8aS)-Decahydro-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethyl]-2(5H)-furanone, ent-19-Hydroxy-8(17),13-labdadien-16,15-olide
Empirical Formula (Hill Notation):
C20H30O3
CAS Number:
Molecular Weight:
318.45
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing