
Amcinonide

Synonym(s):
Amcinonide
Empirical Formula (Hill Notation):
C28H35FO7
CAS Number:
Molecular Weight:
502.57
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing