
α,α-Dimethylphenethyl acetate

Empirical Formula (Hill Notation):
C12H16O2
CAS Number:
Molecular Weight:
192.25
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing