
9-cis-Retinoic acid

Synonym(s):
9-cis-Tretinoin, Alitretinoin
Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing