![7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin ≥95% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/457/098/e3d5a2d0-5cf3-43b6-a14d-84b22de9569f/640/e3d5a2d0-5cf3-43b6-a14d-84b22de9569f.png)
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin
![7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin ≥95% (HPLC), solid](/deepweb/assets/sigmaaldrich/product/structures/457/098/e3d5a2d0-5cf3-43b6-a14d-84b22de9569f/640/e3d5a2d0-5cf3-43b6-a14d-84b22de9569f.png)
Synonym(s):
1H-Pyrrole-2,5-dione, 1-(4-(7-(diethylamino)-4-methyl-2-oxo-2H-1-benzopyran-3-yl)phenyl)-, CPM
Empirical Formula (Hill Notation):
C24H22N2O4
CAS Number:
Molecular Weight:
402.44
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-2 of 2