4-Hydroxy-L-isoleucine
Synonym(s):
(2S,3R)-2-Amino-4-hydroxy-3-methylpentanoic acid
Empirical Formula (Hill Notation):
C6H13NO3
CAS Number:
Molecular Weight:
147.17
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Optical Isomer
Amino Acid
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing